Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007707.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007707.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007707.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007707.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007707.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007707.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155734.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155734.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155734.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155734.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155734.hg.1 |
TC0300007707.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155735.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155735.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155735.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155735.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155735.hg.1 |
TC0300007707.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007707.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007707.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007707.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007707.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007707.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007707.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155733.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155733.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155733.hg.1 |
ENST00000472514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155733.hg.1 |
NR_046575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155733.hg.1 |
TC0300007707.hg.1 |
Clariom D TC |