Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161500.hg.1 |
TC0300008661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300161501.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161501.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161501.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161501.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161501.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161501.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161501.hg.1 |
TC0300008661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300161502.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161502.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161502.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161502.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161502.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161502.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161502.hg.1 |
TC0300008661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008661.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161495.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161495.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161495.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161495.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161495.hg.1 |
TC0300008661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
ENST00000500525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161499.hg.1 |
TC0300008661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300161494.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161494.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161494.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161494.hg.1 |
ENST00000500232.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161494.hg.1 |
ENST00000502278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161494.hg.1 |
NR_046383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161494.hg.1 |
TC0300008661.hg.1 |
Clariom D TC |