This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332301_m1
See other ENC1 GE Assays ›
Gene Symbol:
ENC1
Gene Name:
ectodermal-neural cortex 1
Gene Aliases:
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10
Chromosome Location:
Chr.5: 74627406 - 74641424 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332301_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ENC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046318.1 1 - 2 1113 94
GenBank mRNA DA785554.1 - 1 - 2 417 94
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA168651.1 - 1 - 2 417 Chr. 5 - 74629859 - 74640728 [-] on Build GRCh38 94
GenBankmRNA DA497062.1 - 2 - 3 325 Chr. 5 - 74629763 - 74640728 [-] on Build GRCh38 94
GenBankmRNA AA631247.1 - 1 - 2 210 Chr. 5 - 74629942 - 74640522 [-] on Build GRCh38 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 1331 LINC01331 -90.4kb Sense Chr. 5 - 74369376 - 74536976 on NCBI Build GRCh38
Downstream G elongation factor mitochondrial 2 GFM2 EF-G2mt,
EFG2,
MRRF2,
MST027,
MSTP027,
RRF2,
RRF2mt,
hEFG2,
mEF-G 2
79.8kb Sense Chr. 5 - 74721204 - 74767371 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
proteasomal ubiquitin-independent protein catabolic process
positive regulation of neuron projection development
protein ubiquitination
negative regulation of translation
actin binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0