Assay Details
Gene Symbol: MIR1208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031613.1 | 1 | 28 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Pvt1 oncogene (non-protein coding) | PVT1 |
LINC00079, MYC, NCRNA00079, onco-lncRNA-100 |
-48.9kb | Sense | Chr. 8 - 127794533 - 128101253 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 824 | LINC00824 | LINC01263 | 255.1kb | Anti-sense | Chr. 8 - 128405269 - 128564679 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800008862.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000756.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463181 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463184 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463155 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463207 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463206 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463170 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16134592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463202 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745299 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463197 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463165 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463210 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463195 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463174 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463190 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463208 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463152 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463200 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745292 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463157 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463188 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463159 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463177 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463207 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463195 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463152 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463188 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463206 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463155 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463184 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16134592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745299 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463181 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463170 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463165 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463202 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463159 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463210 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463197 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463157 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463177 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463174 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745292 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26745304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463200 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463190 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06463208 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08010119.hg.1 | ENST00000408334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08010119.hg.1 | NR_031613.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08010119.hg.1 | TC08000756.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0800008862.hg.1 | ENST00000408334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008862.hg.1 | NR_031613.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160653.hg.1 | ENST00000408334.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160653.hg.1 | NR_031613.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160653.hg.1 | ENST00000408334.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160653.hg.1 | NR_031613.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160653.hg.1 | TC0800008862.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800008862.hg.1 | ENST00000408334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008862.hg.1 | NR_031613.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3310673 | NR_031613.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3310673 | NR_031613.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310673 | NR_031613.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310673 | NR_031613.1 | human_transcriptome |
