Assay Details
Gene Symbol: MIR216A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029629.1 | 1 | 56 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105374690 | LOC105374690 | Anti-sense | Chr. 2 - 55945824 - 56177554 on NCBI Build GRCh38 | ||
Downstream | microRNA 216b | MIR216B |
MIRN216B, mir-216b |
11.7kb | Sense | Chr. 2 - 56000714 - 56000795 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200012662.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001871.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197051 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197047 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277720 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197066 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197044 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197055 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277721 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277766 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277713 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197057 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277736 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277728 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277726 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197037 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277722 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197021 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197016 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277763 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197019 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277730 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197052 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197023 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197067 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197069 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277732 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277723 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277763 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197037 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277720 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277723 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197021 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277732 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197044 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197051 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197055 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197069 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197052 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277721 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277730 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277736 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197067 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277766 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197023 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197019 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277722 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197047 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277726 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197066 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197057 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277728 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277713 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21277741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01197016 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.1 ST Array | 8052374 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0200012662.hg.1 | ENST00000385063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012662.hg.1 | NR_029629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02029933.hg.1 | ENST00000385063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02029933.hg.1 | NR_029629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02029933.hg.1 | TC02001871.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0200012662.hg.1 | ENST00000385063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012662.hg.1 | NR_029629.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200192447.hg.1 | ENST00000385063.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200192447.hg.1 | NR_029629.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200192447.hg.1 | ENST00000385063.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200192447.hg.1 | NR_029629.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200192447.hg.1 | TC0200012662.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013666 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013666 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3311045 | NR_029629.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3311045 | NR_029629.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3311045 | NR_029629.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3311045 | NR_029629.1 | human_transcriptome |
