This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04330779_g1
See other SLC28A3 GE Assays ›
Gene Symbol:
SLC28A3
Gene Name:
solute carrier family 28 member 3
Gene Aliases:
CNT3
Chromosome Location:
Chr.9: 84273123 - 84368634 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04330779_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC28A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022127.2 NP_071410.1 2 72 89
XM_011518906.2 XP_011517208.1 2 208 89
GenBank mRNA AF305210.1 - 2 72 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900010591.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001286.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF305210.1,NM_022127.2,XM_011518906.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Detect Genomic DNA Homo sapiens Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC28A3

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
pyrimidine-containing compound transmembrane transport
purine nucleobase transmembrane transport
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09016094.hg.1 ENST00000376238.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016094.hg.1 ENST00000495823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016094.hg.1 ENST00000537648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016094.hg.1 NM_001199633.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016094.hg.1 NM_022127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016094.hg.1 NR_037638.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016094.hg.1 TC09001286.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09016100.hg.1 NM_022127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016100.hg.1 TC09001286.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09016093.hg.1 ENST00000376238.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016093.hg.1 ENST00000495823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016093.hg.1 ENST00000537648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016093.hg.1 NM_001199633.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016093.hg.1 NM_022127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016093.hg.1 NR_037638.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016093.hg.1 TC09001286.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 ENST00000376238.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 ENST00000495823.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 NM_001199633.1 Refseq
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 NM_022127.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 NR_037638.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 ENST00000376238.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 ENST00000495823.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 NM_001199633.1 Refseq
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 NM_022127.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 NR_037638.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172288.hg.1 TC0900010591.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 ENST00000376238.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 ENST00000495823.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 NM_001199633.1 Refseq
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 NM_022127.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 NR_037638.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 ENST00000376238.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 ENST00000495823.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 NM_001199633.1 Refseq
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 NM_022127.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 NR_037638.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172290.hg.1 TC0900010591.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 ENST00000376238.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 ENST00000495823.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 NM_001199633.1 Refseq
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 NM_022127.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 NR_037638.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 ENST00000376238.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 ENST00000495823.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 NM_001199633.1 Refseq
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 NM_022127.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 NR_037638.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172289.hg.1 TC0900010591.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900172295.hg.1 NM_022127.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172295.hg.1 NM_022127.2 Refseq
Affymetrix Human Clariom D Assay PSR0900172295.hg.1 TC0900010591.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0