This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04275663_s1
See other PRR7-AS1 GE Assays ›
Gene Symbol:
PRR7-AS1
Gene Name:
PRR7 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 177437889 - 177447944 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04275663_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRR7-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038915.1 4 724 83
NR_038916.1 3 525 83
GenBank mRNA BM083785.1 - 4 480 83
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI008816.1 - 223 Chr. - - [-] on Build 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream G protein-coupled receptor kinase 6 GRK6 GPRK6 Anti-sense Chr. 5 - 177426496 - 177442849 on NCBI Build GRCh38
Downstream proline rich 7 (synaptic) PRR7 7.5kb Anti-sense Chr. 5 - 177445936 - 177456701 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500012973.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05002106.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3679876 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 45921 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3679876 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3679876 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1560448_PM_at - -
Affymetrix Human Transcriptome Array 2.0 PSR05029037.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05029037.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05029037.hg.1 NR_038916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05029037.hg.1 TC05002106.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000506465.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000506465.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000511565.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000511565.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000511565.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000514846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000514846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000514846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 ENST00000514846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05029036.hg.1 NR_038915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05029036.hg.1 NR_038916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05029036.hg.1 TC05002106.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500190433.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Clariom D Assay PSR0500190433.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay PSR0500190433.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay PSR0500190433.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Clariom D Assay PSR0500190433.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay PSR0500190433.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay PSR0500190433.hg.1 TC0500012973.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500190432.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay PSR0500190432.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay PSR0500190432.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay PSR0500190432.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay PSR0500190432.hg.1 TC0500012973.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000425316.3 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000506465.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000506465.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000511565.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000511565.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000511565.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000514846.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000514846.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000514846.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 ENST00000514846.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038915.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038916.1 Refseq
Affymetrix Human Clariom D Assay TC0500012973.hg.1 NR_038916.1 Refseq
Human Genome U133 Plus 2.0 Array 1560448_at - -
Human Genome U133 Plus 2.0 Array 1560448_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560448_at6 - -
Human Genome U95 Set 71393_PM_at - -
Human Genome U95 Set 71393_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A027743 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A000174 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A027743 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A000174 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header