This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04274922_s1
See other OR10V2P GE Assays ›
Gene Symbol:
OR10V2P
Gene Name:
olfactory receptor family 10 subfamily V member 2 pseudogene
Gene Aliases:
-
Chromosome Location:
Chr.11: 59748812 - 59749581 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04274922_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OR10V2P
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045005.1 1 418 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream olfactory receptor family 10 subfamily V member 1 OR10V1 OR11-256 -35.0kb Anti-sense Chr. 11 - 59712916 - 59713845 on NCBI Build GRCh38
Downstream syntaxin 3 STX3 STX3A 5.5kb Sense Chr. 11 - 59755059 - 59805882 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100007752.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000500.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100007752.hg.1 ENST00000525296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007752.hg.1 NR_045005.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11005948.hg.1 NR_045005.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11005948.hg.1 TC11000500.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100154599.hg.1 ENST00000525296.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100154599.hg.1 NR_045005.1 Refseq
Affymetrix Human Clariom D Assay PSR1100154599.hg.1 ENST00000525296.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100154599.hg.1 NR_045005.1 Refseq
Affymetrix Human Clariom D Assay PSR1100154599.hg.1 TC1100007752.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100007752.hg.1 ENST00000525296.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007752.hg.1 NR_045005.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0