Assay Details
Gene Symbol: HMGN3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040671.1 | 1 - 2 | 251 | 92 | ||
GenBank mRNA | BC042096.1 | - | 1 - 2 | 92 | ||
DA046558.1 | - | 1 - 2 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000755.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25041602 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15833616 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25041606 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 585700 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 13377 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37676896 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1559404_PM_a_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1559403_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06009046.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06009046.hg.1 | TC06000755.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06009045.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06009045.hg.1 | TC06000755.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008605.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159791.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159791.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159791.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159791.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159791.hg.1 | TC0600008605.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600159789.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159789.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159789.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159789.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159789.hg.1 | TC0600008605.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600159786.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159786.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159786.hg.1 | TC0600008605.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600159792.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159792.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159792.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159792.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159792.hg.1 | TC0600008605.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600159790.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159790.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159790.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159790.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159790.hg.1 | TC0600008605.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600159788.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159788.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159788.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159788.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159788.hg.1 | TC0600008605.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600159787.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159787.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159787.hg.1 | ENST00000604516.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159787.hg.1 | NR_040671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159787.hg.1 | TC0600008605.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1559403_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1559404_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1559404_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1559403_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559404_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559403_at2 | - | - |
Human Genome U95 Set | 52738_PM_at | - | - |
Human Genome U95 Set | 52738_at | - | - |
Human Genome U95 Set | 86625_at | - | - |
Human Genome U95 Set | 86625_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 180737 | - | - |
