This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04274688_s1
See other MIR4753 GE Assays ›
Gene Symbol:
MIR4753
Gene Name:
microRNA 4753
Gene Aliases:
mir-4753
Chromosome Location:
Chr.1: 235190034 - 235190116 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04274688_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR4753
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_039908.1 1 38 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream AT-rich interaction domain 4B ARID4B BCAA,
BRCAA1,
RBBP1L1,
RBP1L1,
SAP180
Sense Chr. 1 - 235166895 - 235328240 on NCBI Build GRCh38
Downstream geranylgeranyl diphosphate synthase 1 GGPS1 GGPPS,
GGPPS1
138.3kb Anti-sense Chr. 1 - 235328438 - 235344532 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0100017821.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003993.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100017821.hg.1 ENST00000585119.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017821.hg.1 NR_039908.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01060447.hg.1 NR_039908.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01060447.hg.1 TC01003993.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0100017821.hg.1 ENST00000585119.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017821.hg.1 NR_039908.1 Refseq
Affymetrix Human Clariom D Assay PSR0100234490.hg.1 ENST00000585119.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100234490.hg.1 NR_039908.1 Refseq
Affymetrix Human Clariom D Assay PSR0100234490.hg.1 ENST00000585119.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100234490.hg.1 NR_039908.1 Refseq
Affymetrix Human Clariom D Assay PSR0100234490.hg.1 TC0100017821.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0