This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273577_s1
See other YY1P2 GE Assays ›
Gene Symbol:
YY1P2
Gene Name:
YY1 transcription factor pseudogene 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 138897324 - 138899174 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273577_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YY1P2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033658.1 1 918 103
GenBank mRNA BC042175.1 - 1 915 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream neurexophilin 2 NXPH2 NPH2 -117.1kb Anti-sense Chr. 2 - 138669157 - 138780241 on NCBI Build GRCh38
Downstream LDL receptor related protein 1B LRP1B LRP-1B,
LRP-DIT,
LRPDIT
1.3mb Anti-sense Chr. 2 - 140231423 - 142132463 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200009495.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000896.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511522 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635977 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635949 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635967 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635947 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511534 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336194 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635975 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511489 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511535 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636032 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635952 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336211 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635963 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511494 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511523 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635935 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511573 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635957 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336190 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511488 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635958 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336197 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511516 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636029 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336205 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635989 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635970 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635946 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511536 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635959 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636033 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635965 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336196 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511567 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635984 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511549 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511557 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336207 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511548 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511529 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511540 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511561 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511560 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511564 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635962 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636037 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635973 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635978 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635939 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636026 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636015 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635993 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636031 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635971 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511493 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636034 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635995 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636016 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336188 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511510 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511559 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511537 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21636019 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635961 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336206 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511491 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511546 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15336210 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635955 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511558 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635992 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P01511574 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P21635960 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125203 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 38366 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3406563 - -
Agilent Human 1B Microarray G4111A A_32_P820198 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009495.hg.1 ENST00000438766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009495.hg.1 ENST00000438766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009495.hg.1 NR_033658.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009495.hg.1 NR_033658.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02013272.hg.1 NR_033658.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02013272.hg.1 TC02000896.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200166431.hg.1 ENST00000438766.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200166431.hg.1 NR_033658.1 Refseq
Affymetrix Human Clariom D Assay PSR0200166431.hg.1 ENST00000438766.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200166431.hg.1 NR_033658.1 Refseq
Affymetrix Human Clariom D Assay PSR0200166431.hg.1 TC0200009495.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200166432.hg.1 NR_033658.1 Refseq
Affymetrix Human Clariom D Assay PSR0200166432.hg.1 NR_033658.1 Refseq
Affymetrix Human Clariom D Assay PSR0200166432.hg.1 TC0200009495.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200009495.hg.1 ENST00000438766.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009495.hg.1 ENST00000438766.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009495.hg.1 NR_033658.1 Refseq
Affymetrix Human Clariom D Assay TC0200009495.hg.1 NR_033658.1 Refseq
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A016426 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A016426 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3284530 NR_033658.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3284530 BC042175.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0