Assay Details
Gene Symbol: MIR1471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031717.1 | 1 | 29 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | COP9 signalosome subunit 7B | COPS7B |
CSN7B, SGN7b |
-83.0kb | Anti-sense | Chr. 2 - 231781671 - 231809253 on NCBI Build GRCh38 |
Downstream | natriuretic peptide C | NPPC |
CNP, CNP2 |
29.8kb | Sense | Chr. 2 - 231922094 - 231926403 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200016044.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002868.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960781 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087524 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960798 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960822 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087523 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087512 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087514 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087520 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960824 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960815 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960818 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087494 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960797 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087490 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087524 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087490 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960798 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087512 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087514 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087520 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960822 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087494 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087523 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960818 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960797 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22087503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960815 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960824 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960781 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01960816 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016044.hg.1 | ENST00000410314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016044.hg.1 | NR_031717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02046240.hg.1 | ENST00000410314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02046240.hg.1 | NR_031717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02046240.hg.1 | TC02002868.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0200217876.hg.1 | ENST00000410314.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217876.hg.1 | NR_031717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217876.hg.1 | ENST00000410314.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217876.hg.1 | NR_031717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217876.hg.1 | TC0200016044.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200016044.hg.1 | ENST00000410314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016044.hg.1 | NR_031717.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3308627 | NR_031717.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3308627 | NR_031717.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308627 | NR_031717.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308627 | NR_031717.1 | human_transcriptome |
