Assay Details
Gene Symbol: MIR1301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031570.1 | 1 | 41 | 54 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | proopiomelanocortin | POMC |
ACTH, CLIP, LPH, MSH, NPP, POC |
-159.8kb | Sense | Chr. 2 - 25160853 - 25168851 on NCBI Build GRCh38 |
Downstream | dystrobrevin beta | DTNB | 48.5kb | Sense | Chr. 2 - 25377220 - 25673647 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200012005.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001659.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054981 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133842 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054979 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054953 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054930 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054934 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054933 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133837 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054967 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054938 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054986 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133885 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054971 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133828 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054978 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133874 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054934 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054971 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054938 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133837 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054967 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054981 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054978 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133828 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133885 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15226783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054986 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054933 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054979 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133842 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054953 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054930 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133874 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01054987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21133838 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02026507.hg.1 | ENST00000408518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026507.hg.1 | NR_031570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026507.hg.1 | TC02001659.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200012005.hg.1 | ENST00000408518.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012005.hg.1 | NR_031570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012005.hg.1 | ENST00000408518.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012005.hg.1 | NR_031570.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187327.hg.1 | ENST00000408518.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187327.hg.1 | NR_031570.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187327.hg.1 | ENST00000408518.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187327.hg.1 | NR_031570.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187327.hg.1 | TC0200012005.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3309374 | NR_031570.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3309374 | NR_031570.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309374 | NR_031570.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309374 | NR_031570.1 | human_transcriptome |
