This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273379_s1
See other MIR885 GE Assays ›
Gene Symbol:
MIR885
Gene Name:
microRNA 885
Gene Aliases:
MIRN885, hsa-mir-885, mir-885
Chromosome Location:
Chr.3: 10394489 - 10394562 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273379_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR885
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030614.1 1 31 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 378b MIR378B -64.2kb Anti-sense Chr. 3 - 10330229 - 10330285 on NCBI Build GRCh38
Downstream ATP2B2 intronic transcript 2 ATP2B2-IT2 231.5kb Sense Chr. 3 - 10626015 - 10626807 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0300010243.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001161.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188068 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055693 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188065 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188088 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188061 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055694 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055660 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055656 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055684 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188058 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188051 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055698 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15411879 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055675 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188093 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188089 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055672 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055662 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055690 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188077 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055686 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188075 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188060 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055661 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188091 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055658 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055681 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055657 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188067 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055677 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188056 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055686 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15411879 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055690 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055662 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055657 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188058 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055693 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188068 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055684 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055675 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188077 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055681 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055656 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188060 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055698 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188067 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055661 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188091 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188061 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188088 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188093 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055660 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055694 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055672 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188075 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055677 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188089 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188051 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188065 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22188056 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02055658 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300010243.hg.1 ENST00000401316.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010243.hg.1 NR_030614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03020132.hg.1 ENST00000401316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03020132.hg.1 NR_030614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03020132.hg.1 TC03001161.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0300010243.hg.1 ENST00000401316.2 Ensembl
Affymetrix Human Clariom D Assay TC0300010243.hg.1 NR_030614.1 Refseq
Affymetrix Human Clariom D Assay PSR0300174824.hg.1 ENST00000401316.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300174824.hg.1 NR_030614.1 Refseq
Affymetrix Human Clariom D Assay PSR0300174824.hg.1 ENST00000401316.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300174824.hg.1 NR_030614.1 Refseq
Affymetrix Human Clariom D Assay PSR0300174824.hg.1 TC0300010243.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309329 NR_030614.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309329 NR_030614.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309329 NR_030614.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309329 NR_030614.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0