This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273332_s1
See other MIR624 GE Assays ›
Gene Symbol:
MIR624
Gene Name:
microRNA 624
Gene Aliases:
MIRN624, hsa-mir-624, mir-624
Chromosome Location:
Chr.14: 31014646 - 31014742 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273332_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR624
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030354.1 1 45 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream striatin 3 STRN3 PPP2R6B,
SG2NA
Sense Chr. 14 - 30893799 - 31026528 on NCBI Build GRCh38
Downstream adaptor related protein complex 4 sigma 1 subunit AP4S1 AP47B,
CLA20,
CLAPS4,
CPSQ6,
SPG52
10.4kb Anti-sense Chr. 14 - 31025106 - 31096450 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1400008841.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14001008.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09202360 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29659953 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29659969 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29659959 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09202358 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29659967 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16691089 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16691093 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09202362 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09202368 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16691089 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16691093 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09202368 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09202360 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29659959 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09202358 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29659969 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09202362 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29659953 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29659967 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14011505.hg.1 ENST00000385217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011505.hg.1 NR_030354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14011505.hg.1 TC14001008.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1400008841.hg.1 ENST00000385217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008841.hg.1 NR_030354.1 Refseq
Affymetrix Human Clariom D Assay PSR1400165047.hg.1 ENST00000385217.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400165047.hg.1 NR_030354.1 Refseq
Affymetrix Human Clariom D Assay PSR1400165047.hg.1 ENST00000385217.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400165047.hg.1 NR_030354.1 Refseq
Affymetrix Human Clariom D Assay PSR1400165047.hg.1 TC1400008841.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1400008841.hg.1 ENST00000385217.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008841.hg.1 NR_030354.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308673 NR_030354.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308673 NR_030354.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308673 NR_030354.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308673 NR_030354.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0