This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273326_s1
See other MIR609 GE Assays ›
Gene Symbol:
MIR609
Gene Name:
microRNA 609
Gene Aliases:
MIRN609, hsa-mir-609
Chromosome Location:
Chr.10: 104218789 - 104218883 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273326_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR609
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030340.1 1 45 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SWI5 dependent homologous recombination repair protein 1 SFR1 C10orf78,
MEI5,
MEIR5,
bA373N18.1
-92.4kb Anti-sense Chr. 10 - 104122058 - 104126385 on NCBI Build GRCh38
Downstream glutathione S-transferase omega 1 GSTO1 GSTO 1-1,
GSTTLp28,
HEL-S-21,
P28,
SPG-R
35.3kb Anti-sense Chr. 10 - 104254194 - 104267464 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1000011744.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001639.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837928 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446858 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446844 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837925 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446867 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837910 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837902 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837919 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446855 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837927 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446860 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837898 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837909 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837900 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837922 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837911 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446845 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837915 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837901 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446862 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837926 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837913 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837899 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837917 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837894 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446872 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446856 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446847 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837915 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837910 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837898 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837901 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837925 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446860 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446856 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837927 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837913 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446867 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446855 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837917 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837926 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446858 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446872 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837894 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837919 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446862 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837928 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837900 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837909 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837922 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837911 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446844 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837899 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446845 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837902 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07446847 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27837924 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011744.hg.1 ENST00000385298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011744.hg.1 NR_030340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10021540.hg.1 ENST00000385298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10021540.hg.1 NR_030340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10021540.hg.1 TC10001639.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000180474.hg.1 ENST00000385298.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180474.hg.1 NR_030340.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180474.hg.1 ENST00000385298.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180474.hg.1 NR_030340.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180474.hg.1 TC1000011744.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000011744.hg.1 ENST00000385298.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011744.hg.1 NR_030340.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309399 NR_030340.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309399 NR_030340.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309399 NR_030340.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309399 NR_030340.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0