This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273250_s1
See other MIR126 GE Assays ›
Gene Symbol:
MIR126
Gene Name:
microRNA 126
Gene Aliases:
MIRN126, miRNA126, mir-126
Chromosome Location:
Chr.9: 136670602 - 136670686 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273250_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR126
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029695.1 1 44 75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX381731.2 - 1 416 Chr. 9 - 136670229 - 136672009 [+] on Build GRCh38 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101928612 HSPC324 -10.1kb Anti-sense Chr. 9 - 136648610 - 136660421 on NCBI Build GRCh38
Downstream 1-acylglycerol-3-phosphate O-acyltransferase 2 AGPAT2 1-AGPAT2,
BSCL,
BSCL1,
LPAAB,
LPAAT-beta
2.5kb Anti-sense Chr. 9 - 136673143 - 136687459 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0900009227.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000818.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009758 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009753 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009742 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009762 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360936 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360955 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009772 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009768 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360964 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009761 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360960 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360929 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009760 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009759 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009748 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009755 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009741 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360915 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009745 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360957 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009771 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009744 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360947 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009752 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360970 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360926 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009747 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009750 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360916 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009737 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009749 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009748 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009741 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009753 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009745 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009747 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009742 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009752 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009768 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009749 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360970 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360915 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009760 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009761 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360916 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009755 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009744 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360957 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360929 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009762 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360926 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009758 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360960 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360947 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009737 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360936 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360964 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009759 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009750 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27360955 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009771 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07009772 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 8159371 - -
Affymetrix Human Transcriptome Array 2.0 PSR09010598.hg.1 ENST00000362291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09010598.hg.1 NR_029695.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09010598.hg.1 TC09000818.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0900009227.hg.1 ENST00000362291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009227.hg.1 NR_029695.1 Refseq
Affymetrix Human Clariom D Assay TC0900009227.hg.1 ENST00000362291.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009227.hg.1 NR_029695.1 Refseq
Affymetrix Human Clariom D Assay PSR0900162901.hg.1 ENST00000362291.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900162901.hg.1 NR_029695.1 Refseq
Affymetrix Human Clariom D Assay PSR0900162901.hg.1 ENST00000362291.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900162901.hg.1 NR_029695.1 Refseq
Affymetrix Human Clariom D Assay PSR0900162901.hg.1 TC0900009227.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308073 NR_029695.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3308073 CR600602.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3308073 CR609845.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3308073 CR593160.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3308073 CR593184.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308073 NR_029695.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308073 CR600602.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308073 CR609845.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308073 CR593160.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308073 CR593184.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308073 NR_029695.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308073 CR600602.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308073 CR609845.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308073 CR593160.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308073 CR593184.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308073 NR_029695.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308073 CR600602.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308073 CR609845.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308073 CR593160.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308073 CR593184.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0