This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04260267_s1
See other GPR183 GE Assays ›
Gene Symbol:
GPR183
Gene Name:
G protein-coupled receptor 183
Gene Aliases:
EBI2
Chromosome Location:
Chr.13: 99294273 - 99307495 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04260267_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR183
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004951.4 NP_004942.1 2 517 152
XM_017020405.1 XP_016875894.1 2 460 152
GenBank mRNA AK223390.1 - 2 421 152
AK292091.1 - 2 441 152
AK313443.1 - 2 517 152
BC020752.1 - 2 421 4722289 152
L08177.1 - 1 376 152

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR183

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
B cell activation involved in immune response
mature B cell differentiation involved in immune response
dendritic cell chemotaxis
immune response
humoral immune response
G-protein coupled receptor signaling pathway
T cell chemotaxis
osteoclast differentiation
leukocyte chemotaxis
positive regulation of B cell proliferation
dendritic cell homeostasis
T follicular helper cell differentiation
positive regulation of ERK1 and ERK2 cascade
regulation of astrocyte chemotaxis
G-protein coupled receptor activity
oxysterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0