This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04260260_s1
See other GPR183 GE Assays ›
Gene Symbol:
GPR183
Gene Name:
G protein-coupled receptor 183
Gene Aliases:
EBI2
Chromosome Location:
Chr.13: 99294273 - 99307495 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04260260_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR183
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004951.4 NP_004942.1 2 1121 79
XM_017020405.1 XP_016875894.1 2 1064 79
GenBank mRNA AK223390.1 - 2 1025 79
AK292091.1 - 2 1045 79
BC020752.1 - 2 1025 4722289 79
L08177.1 - 1 980 79

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR183

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
B cell activation involved in immune response
mature B cell differentiation involved in immune response
dendritic cell chemotaxis
immune response
humoral immune response
G-protein coupled receptor signaling pathway
T cell chemotaxis
osteoclast differentiation
leukocyte chemotaxis
positive regulation of B cell proliferation
dendritic cell homeostasis
T follicular helper cell differentiation
positive regulation of ERK1 and ERK2 cascade
regulation of astrocyte chemotaxis
G-protein coupled receptor activity
oxysterol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300009598.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009598.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009598.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009598.hg.1 NM_004951.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009598.hg.1 NM_004951.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009598.hg.1 NM_004951.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009598.hg.1 NM_004951.4 Refseq
Affymetrix Human Clariom D Assay TC1300009598.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009598.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009598.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009598.hg.1 NM_004951.4 Refseq
Affymetrix Human Clariom D Assay TC1300009598.hg.1 NM_004951.4 Refseq
Affymetrix Human Clariom D Assay TC1300009598.hg.1 NM_004951.4 Refseq
Affymetrix Human Clariom D Assay TC1300009598.hg.1 NM_004951.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0