Assay Details
Gene Symbol: LOC115110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037844.2 | 5 | 2798 | 97 | ||
GenBank mRNA | AK128532.1 | - | 5 | 2798 | 97 | |
AL359943.1 | - | 1 | 1851 | 97 | ||
CR749717.1 | - | 1 | 1851 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hes family bHLH transcription factor 5 | HES5 | bHLHb38 | -20.7kb | Sense | Chr. 1 - 2528745 - 2530554 on NCBI Build GRCh38 |
Downstream | TNF receptor superfamily member 14 | TNFRSF14 |
ATAR, CD270, HVEA, HVEM, LIGHTR, TR2 |
2.2kb | Anti-sense | Chr. 1 - 2555767 - 2565622 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | ENST00000424215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | ENST00000424215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | ENST00000424215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | ENST00000424215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | ENST00000456687.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | ENST00000456687.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | ENST00000456687.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | ENST00000456687.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | ENST00000424215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | ENST00000424215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | ENST00000424215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | ENST00000424215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | ENST00000456687.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | ENST00000456687.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | ENST00000456687.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | ENST00000456687.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | NR_121638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018163.hg.1 | NR_121638.1 | Refseq |
