This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04232435_m1
See other C1QTNF3-AMACR GE Assays ›
Gene Symbol:
C1QTNF3-AMACR
Gene Name:
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases:
-
Chromosome Location:
Chr.5: 33986986 - 34124528 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04232435_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C1QTNF3-AMACR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037951.1 6 - 7 765 86
GenBank mRNA AB209261.1 - 6 - 7 774 86
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA FY213173.1 - 3 - 4 Chr. 5 - 33998689 - 34023938 [+] on Build GRCh38 86
GenBankmRNA FY212965.1 - 3 - 4 Chr. 5 - 33998690 - 34023936 [+] on Build GRCh38 86
GenBankmRNA FY211704.1 - 3 - 4 Chr. 5 - 33998689 - 34020732 [+] on Build GRCh38 86
GenBankmRNA FY211019.1 - 2 - 3 Chr. 5 - 33998689 - 34028797 [+] on Build GRCh38 86
GenBankmRNA FY212525.1 - 2 - 3 Chr. 5 - 33998690 - 34028793 [+] on Build GRCh38 86
GenBankmRNA FY212703.1 - 3 - 4 Chr. 5 - 33998689 - 34023945 [+] on Build GRCh38 86
GenBankmRNA FY211892.1 - 3 - 4 Chr. 5 - 33998689 - 34020739 [+] on Build GRCh38 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream C1q and tumor necrosis factor related protein 3 C1QTNF3 C1ATNF3,
CORCS,
CORS,
CORS-26,
CORS26,
CTRP3
Sense Chr. 5 - 34017858 - 34043266 on NCBI Build GRCh38
Downstream integral membrane glycoprotein-like LOC646652 168.8kb Anti-sense Chr. 5 - 34189581 - 34194204 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500013296.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay PSR0500195336.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay PSR0500195336.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay PSR0500195336.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay PSR0500195336.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay PSR0500195336.hg.1 TC0500013296.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 ENST00000382079.3 Ensembl
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq
Affymetrix Human Clariom D Assay TC0500013296.hg.1 NR_037951.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0