This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04232274_m1
See other AP4B1-AS1 GE Assays ›
Gene Symbol:
AP4B1-AS1
Gene Name:
AP4B1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 113812612 - 113901237 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04232274_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AP4B1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037864.1 1 - 2 87 108
GenBank mRNA AK123199.1 - 1 - 2 87 107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA136781.1 - 1 - 2 87 Chr. 1 - 113856635 - 113900053 [+] on Build GRCh38 107

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream round spermatid basic protein 1 RSBN1 ROSBIN -44.2kb Anti-sense Chr. 1 - 113761832 - 113812476 on NCBI Build GRCh38
Downstream BCL2 like 15 BCL2L15 Bfk,
C1orf178
19.2kb Anti-sense Chr. 1 - 113876814 - 113887547 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100009477.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001001.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01015963.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01015963.hg.1 NR_037864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01015963.hg.1 TC01001001.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01015962.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01015962.hg.1 NR_037864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01015962.hg.1 TC01001001.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000418238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000418238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000418238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000448199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 ENST00000448199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Clariom D Assay PSR0100168300.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100168300.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay PSR0100168300.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100168300.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay PSR0100168300.hg.1 TC0100009477.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000418238.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000418238.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000418238.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000429398.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000448199.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 ENST00000448199.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Clariom D Assay TC0100009477.hg.1 NR_125965.1 Refseq
Affymetrix Human Clariom D Assay PSR0100168301.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100168301.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay PSR0100168301.hg.1 ENST00000419536.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100168301.hg.1 NR_037864.1 Refseq
Affymetrix Human Clariom D Assay PSR0100168301.hg.1 TC0100009477.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0