This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231863_s1
See other MIR4251 GE Assays ›
Gene Symbol:
MIR4251
Gene Name:
microRNA 4251
Gene Aliases:
-
Chromosome Location:
Chr.1: 3127975 - 3128035 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231863_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR4251
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036215.1 1 32 56

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 982 LINC00982 -60.3kb Anti-sense Chr. 1 - 3059617 - 3067725 on NCBI Build GRCh38
Downstream uncharacterized LOC105378604 LOC105378604 178.6kb Anti-sense Chr. 1 - 3306636 - 3327974 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0100006605.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000068.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01001076.hg.1 NR_036215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001076.hg.1 TC01000068.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100145709.hg.1 NR_036215.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145709.hg.1 NR_036215.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145709.hg.1 TC0100006605.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0