Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010855.hg.1 |
ENST00000365731.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010855.hg.1 |
NR_030308.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175710.hg.1 |
ENST00000365731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175710.hg.1 |
NR_030308.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175710.hg.1 |
ENST00000365731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175710.hg.1 |
NR_030308.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175710.hg.1 |
TC0500010855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010855.hg.1 |
ENST00000365731.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010855.hg.1 |
NR_030308.1 |
Refseq |