Assay Details
Gene Symbol: MIR551B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030294.1 | 1 | 44 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
EGF like and EMI domain containing 1, pseudogene |
EGFEM1P |
C3orf50, NCRNA00259 |
Sense | Chr. 3 - 168249522 - 168830599 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC100507661 | LOC100507661 | 350.0kb | Sense | Chr. 3 - 168901945 - 168921996 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300009425.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000900.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890631 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750599 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890662 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890638 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890630 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890628 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750586 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750609 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890627 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890654 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750600 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750634 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890666 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750608 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890666 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890662 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750600 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890631 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750586 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750609 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890630 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890627 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750608 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890654 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890638 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750599 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750634 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890628 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22890639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02750632 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03015817.hg.1 | ENST00000384984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015817.hg.1 | NR_030294.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015817.hg.1 | TC03000900.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300009425.hg.1 | ENST00000384984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009425.hg.1 | NR_030294.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009425.hg.1 | ENST00000384984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009425.hg.1 | NR_030294.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167763.hg.1 | ENST00000384984.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167763.hg.1 | NR_030294.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167763.hg.1 | ENST00000384984.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300167763.hg.1 | NR_030294.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300167763.hg.1 | TC0300009425.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3311090 | NR_030294.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3311090 | NR_030294.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3311090 | NR_030294.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3311090 | NR_030294.1 | human_transcriptome |
