This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231653_s1
See other MIR504 GE Assays ›
Gene Symbol:
MIR504
Gene Name:
microRNA 504
Gene Aliases:
MIRN504, hsa-mir-504, mir-504
Chromosome Location:
Chr.X: 138667711 - 138667793 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231653_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR504
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030229.1 1 43 57

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 889 LINC00889 -50.1kb Sense Chr. X - 138614731 - 138617638 on NCBI Build GRCh38
Downstream FGF13 antisense RNA 1 FGF13-AS1 44.3kb Anti-sense Chr. X - 138712107 - 138716605 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0X00010955.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001426.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717305 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717294 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717292 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717287 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717281 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717301 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717304 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984052 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984057 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717307 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717290 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984042 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717302 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717289 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984041 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717282 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717298 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717288 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717313 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984066 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717310 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717293 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717284 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717297 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984037 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717300 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717306 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717312 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984042 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984037 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717297 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717290 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717305 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717287 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984041 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717289 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717306 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717300 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717312 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717281 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984057 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717310 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717292 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717302 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717284 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984066 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717298 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717307 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11984052 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717313 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717282 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717304 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717288 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717294 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717301 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32717293 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 8175455 - -
Affymetrix Human Transcriptome Array 2.0 TC0X00010955.hg.1 ENST00000385065.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010955.hg.1 NR_030229.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X018918.hg.1 ENST00000385065.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X018918.hg.1 NR_030229.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X018918.hg.1 TC0X001426.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0X00010955.hg.1 ENST00000385065.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00010955.hg.1 NR_030229.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00174467.hg.1 ENST00000385065.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00174467.hg.1 NR_030229.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00174467.hg.1 ENST00000385065.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00174467.hg.1 NR_030229.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00174467.hg.1 TC0X00010955.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309869 NR_030229.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309869 NR_030229.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0