This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231519_s1
See other MIR134 GE Assays ›
Gene Symbol:
MIR134
Gene Name:
microRNA 134
Gene Aliases:
MIRN134, mir-134
Chromosome Location:
Chr.14: 101054687 - 101054759 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231519_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR134
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029698.1 1 38 65

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1400008297.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000750.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16735000 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16735000 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 7976844 - -
Affymetrix Human Transcriptome Array 2.0 PSR14008215.hg.1 ENST00000385258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14008215.hg.1 NR_029698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14008215.hg.1 TC14000750.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1400008297.hg.1 ENST00000385258.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008297.hg.1 NR_029698.1 Refseq
Affymetrix Human Clariom D Assay PSR1400159087.hg.1 ENST00000385258.3 Ensembl
Affymetrix Human Clariom D Assay PSR1400159087.hg.1 NR_029698.1 Refseq
Affymetrix Human Clariom D Assay PSR1400159087.hg.1 ENST00000385258.3 Ensembl
Affymetrix Human Clariom D Assay PSR1400159087.hg.1 NR_029698.1 Refseq
Affymetrix Human Clariom D Assay PSR1400159087.hg.1 TC1400008297.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1400008297.hg.1 ENST00000385258.3 Ensembl
Affymetrix Human Clariom D Assay TC1400008297.hg.1 NR_029698.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308043 NR_029698.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308043 NR_029698.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308043 NR_029698.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308043 NR_029698.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0