This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04195228_s1
See other S100A7A GE Assays ›
Gene Symbol:
S100A7A
Gene Name:
S100 calcium binding protein A7A
Gene Aliases:
NICE-2, S100A15, S100A7L1, S100A7f
Chromosome Location:
Chr.1: 153416524 - 153423225 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04195228_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100A7A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_176823.3 NP_789793.1 3 3453 89
GenBank mRNA AJ243672.2 - 1 1582 89
AY189118.1 - 3 3520 89
BC040254.1 - 1 1263 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100A7A

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

calcium ion binding
protein self-association

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573305 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573341 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516035 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15130030 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573308 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573354 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573326 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573342 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516050 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516047 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516051 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516054 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573389 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15129993 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516046 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516076 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573290 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15129998 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516061 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573298 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516044 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516059 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516048 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516043 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573382 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573338 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516073 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516081 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573357 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516037 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573361 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516041 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573388 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573366 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516078 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15129986 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573364 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573355 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573374 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15130032 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516039 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00516077 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573340 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573363 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20573368 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 64697 - -
Agilent SurePrint G3 Human CGH Microarray 8x60K 27288 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 49034 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 259157 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 255323 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 26755 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 122567 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 212469 AF243433.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 212469 BC021021.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 212469 BC073152.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 212469 BC092471.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 212469 CR618798.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 212469 NM_032565.2 refseq_mrna_human_NM
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P280274 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3393504 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P280274 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 22102 - -
Agilent Human Genome CGH Microarray 44B A_14_P106240 - -
Agilent Human Genome CGH Microarray 44A A_14_P106240 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 57278 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 62523 - -
Agilent Human Genome CGH + SNP Microarray A_16_P30309467 - -
Agilent Human Genome CGH + SNP Microarray A_14_P106240 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P280274 - -
SurePrint G3 Human GE Microarray G4851A A_24_P280274 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573290 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573298 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516047 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516041 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573326 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573308 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573293 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516043 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516046 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129998 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516048 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516032 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516038 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516035 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129997 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129993 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129986 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573305 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516037 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573324 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129993 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573326 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573293 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516035 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573308 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573290 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516046 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516032 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516041 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129986 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516037 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573305 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573324 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516047 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516048 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20573298 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129998 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129997 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516038 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00516043 - -
Whole Human Genome Oligo Microarray G4112A A_24_P280274 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P106240 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P106240 - -
Agilent Human Genome CGH Microarray 4x44K A_14_P106240 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P280274 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 50536 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 23997 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P106240 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P30309543 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10328367 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516050 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516061 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516088 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516074 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516080 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516037 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516039 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516097 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15130045 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516083 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15129993 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516048 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516058 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516046 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516051 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516078 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516052 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516034 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15130030 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516094 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516092 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516038 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516086 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516057 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516071 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516059 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516054 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516076 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516068 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516090 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516075 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516055 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516066 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00516032 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P106240 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P30309543 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NM_176823_at - -
Affymetrix Human Gene 1.0 ST Array 7905580 - -
Affymetrix Human Gene 1.0 ST Array 7905579 - -
Affymetrix Human Gene 1.0 ST Array 7905577 - -
Affymetrix Human Gene 1.1 ST Array 7905576 - -
Affymetrix HT HG-U133+ PM Array Plate 232170_PM_at - -
Affymetrix HT HG-U133+ PM Array Plate 232220_PM_at - -
Affymetrix Human Genome U219 Array 11738062_x_at - -
Affymetrix Human Genome U219 Array 11738062_PM_x_at - -
Affymetrix Human Genome U219 Array 11738063_PM_a_at - -
Affymetrix Human Genome U219 Array 11738063_a_at - -
Human Genome U133 Plus 2.0 Array 232170_PM_at - -
Human Genome U133 Plus 2.0 Array 232220_at - -
Human Genome U133 Plus 2.0 Array 232170_at - -
Human Genome U133 Plus 2.0 Array 232220_PM_at - -
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_176823_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232170_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 232220_at11 - -
Human Genome U133 Set 232220_PM_at - -
Human Genome U133 Set 232220_at - -
Human Genome U133 Set 232170_at - -
Human Genome U133 Set 232170_PM_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A031505 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A031505 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 702561 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1673191 AY189118.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1673191 NM_176823.3 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1673191 AJ243672.2 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_31341192-S AY189118.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_31341192-S NM_176823.3 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_31341192-S AJ243672.2 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_4534-S - -
Illumina human-6 v1.0 expression beadchip 0002360056 - -
Illumina humanRef-8 v1.0 expression beadchip 0002360056 - -
Illumina human-6 v2.0 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_4534 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_4534 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1673191 AY189118.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1673191 NM_176823.3 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1673191 AJ243672.2 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1673191 AY189118.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1673191 NM_176823.3 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1673191 AJ243672.2 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1673191 AY189118.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1673191 NM_176823.3 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1673191 AJ243672.2 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0