Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19_gl000209_random000048.hg.1 |
NM_012313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19_gl000209_random000048.hg.1 |
NM_014513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19_gl000209_random000048.hg.1 |
TC19_gl000209_random000005.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19_gl000209_random000051.hg.1 |
NM_012313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19_gl000209_random000051.hg.1 |
NM_014513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19_gl000209_random000051.hg.1 |
TC19_gl000209_random000005.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
ENST00000291633.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
ENST00000336077.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
ENST00000400864.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
NM_014513.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
ENST00000291633.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
ENST00000336077.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
ENST00000400864.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
NM_014513.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169369.hg.1 |
TC1900008879.hg.1 |
Clariom D TC |