This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04189825_g1
See other DOCK11 GE Assays ›
Gene Symbol:
DOCK11
Gene Name:
dedicator of cytokinesis 11
Gene Aliases:
ACG, ZIZ2, bB128O4.1
Chromosome Location:
Chr.X: 118495664 - 118686162 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04189825_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DOCK11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144658.3 NP_653259.3 46 - 47 5256 97
XM_005262368.3 XP_005262425.1 47 - 48 5540 97
XM_005262369.4 XP_005262426.1 47 - 48 5234 97
XM_011531276.2 XP_011529578.1 47 - 48 5175 97
XM_011531278.2 XP_011529580.1 34 - 35 3661 97
GenBank mRNA AK054649.1 - 7 - 8 938 97
AK056684.1 - 16 - 17 1770 97
AK124735.1 - 41 - 42 4679 97
AK125641.1 - 25 - 26 2768 97
AY692226.1 - 46 - 47 5256 97
BC047713.1 - 12 - 13 1347 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00008228.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000574.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DOCK11

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
blood coagulation
positive regulation of GTPase activity
Rho guanyl-nucleotide exchange factor activity
protein binding
Rho GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X007614.hg.1 ENST00000276202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007614.hg.1 ENST00000276204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007614.hg.1 NM_144658.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007614.hg.1 TC0X000574.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 ENST00000276202.8 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 ENST00000276204.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 ENST00000633080.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 NM_144658.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 ENST00000276202.8 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 ENST00000276204.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 ENST00000633080.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 NM_144658.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00156184.hg.1 TC0X00008228.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0