This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04189688_m1
See other S100A16 GE Assays ›
Gene Symbol:
S100A16
Gene Name:
S100 calcium binding protein A16
Gene Aliases:
AAG13, DT1P1A7, S100F
Chromosome Location:
Chr.1: 153606883 - 153613168 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04189688_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100A16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080388.2 NP_525127.1 1 - 2 218 73
NR_133645.1 1 - 2 218 73
GenBank mRNA AW954845.1 - 1 - 2 194 73
AY221961.1 - 1 - 2 89 73
AY762098.1 - 1 - 2 133 73
BC010541.1 - 1 - 2 88 3458906 73
BC019099.1 - 1 - 2 132 5013080 73
BM739127.1 - 1 - 2 220 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY221961.1, AY762098.1, BC010541.1, BC019099.1, NM_080388.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100A16

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

response to calcium ion
calcium ion binding
protein binding
protein homodimerization activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0