Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033605.hg.1 |
ENST00000377408.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033605.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033605.hg.1 |
ENST00000414642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033605.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033605.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033605.hg.1 |
TC01002180.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000465853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000465853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000465853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033602.hg.1 |
ENST00000377408.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033602.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033602.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033602.hg.1 |
TC01002180.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
ENST00000465853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
ENST00000465853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194595.hg.1 |
TC0100012788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100194592.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194592.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194592.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194592.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194592.hg.1 |
TC0100012788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000377411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000465853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000465853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000465853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
ENST00000466131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012788.hg.1 |
NM_024980.4 |
Refseq |