Assay Details
Gene Symbol: C2CD4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136003.1 | NP_001129475.1 | 1 - 2 | 351 | 54 | |
GenBank mRNA | AK293673.1 | - | 1 - 2 | 351 | 54 | |
DC305608.1 | - | 1 - 2 | 351 | 54 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003235.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293673.1,NM_001136003.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3240244 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1867 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1866 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48211 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3240244 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3240244 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20566381 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20566372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20566384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20566372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20566381 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20566384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00509663 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15128302 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509605 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15128325 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509616 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15128304 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509678 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509601 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509664 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509715 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509707 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509670 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509656 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509676 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509718 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509643 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509634 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509709 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15128323 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509639 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509627 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509711 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509671 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509611 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509625 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509647 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509673 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509705 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509674 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509614 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509622 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509641 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509608 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509620 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509700 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509607 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509623 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509646 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15128319 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509710 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15128337 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509603 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509663 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15128299 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509680 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509716 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00509703 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Genome U219 Array | 11735933_at | - | - |
Affymetrix Human Genome U219 Array | 11735933_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049974.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049974.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049974.hg.1 | TC01003235.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049971.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049971.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049971.hg.1 | TC01003235.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049975.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049975.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049975.hg.1 | TC01003235.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049972.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049972.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049972.hg.1 | TC01003235.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049973.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049973.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01049973.hg.1 | TC01003235.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0100218806.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218806.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218806.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218806.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218806.hg.1 | TC0100015813.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100218807.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218807.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218807.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218807.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218807.hg.1 | TC0100015813.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015813.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218808.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218808.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218808.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218808.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218808.hg.1 | TC0100015813.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100218809.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218809.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218809.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218809.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218809.hg.1 | TC0100015813.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100218810.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218810.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218810.hg.1 | ENST00000454109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218810.hg.1 | NM_001136003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218810.hg.1 | TC0100015813.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052150 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052150 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3248176 | NM_001136003.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3248176 | BC157847.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3248176 | BC171843.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3248176 | NM_001136003.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3248176 | BC157847.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3248176 | BC171843.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248176 | NM_001136003.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248176 | BC157847.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248176 | BC171843.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248176 | NM_001136003.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248176 | BC157847.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248176 | BC171843.1 | human_transcriptome |
