Assay Details
| Gene Symbol: DAOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161812.1 | NP_001155284.1 | 2 - 3 | 252 | 114 | |
| NM_001161814.1 | NP_001155286.1 | 3 - 4 | 446 | 114 | ||
| NM_172370.4 | NP_758958.3 | 2 - 3 | 206 | 114 | ||
| XM_005254042.1 | XP_005254099.1 | 2 - 3 | 404 | 114 | ||
| GenBank mRNA | AY138546.1 | - | 2 - 3 | 179 | 114 | |
| AY138547.1 | - | 3 - 4 | 446 | 114 | ||
| AY170469.2 | - | 3 - 4 | 268 | 114 | ||
| AY170470.2 | - | 3 - 4 | 268 | 114 | ||
| AY223901.1 | - | 2 - 3 | 404 | 114 | ||
| BC121091.1 | - | 2 - 3 | 174 | 40119921 | 114 | |
| DQ343761.1 | - | 2 - 3 | 453 | 114 | ||
| DQ357223.1 | - | 2 - 3 | 198 | 114 | ||
| DQ386869.1 | - | 2 - 3 | 152 | 114 | ||
| DQ386870.1 | - | 2 - 3 | 198 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY170469.2, AY170470.2, DQ343761.1, DQ357223.1, DQ386869.1, DQ386870.1, NM_001161812.1, NM_001161814.1 transcripts.
Set Membership: |
|

DAOA
Genomic Map