This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04185172_m1
See other FER1L6 GE Assays ›
Gene Symbol:
FER1L6
Gene Name:
fer-1 like family member 6
Gene Aliases:
C8ORFK23
Chromosome Location:
Chr.8: 123851987 - 124120061 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04185172_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FER1L6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039112.2 NP_001034201.2 18 - 19 2486 57
XM_006716618.3 XP_006716681.1 18 - 19 2413 57
XM_011517231.2 XP_011515533.1 19 - 20 2452 57
XM_011517232.2 XP_011515534.1 19 - 20 2593 57
XM_011517233.2 XP_011515535.1 18 - 19 2554 57
XM_011517235.2 XP_011515537.1 19 - 20 2453 57
GenBank mRNA AB196633.1 - 18 - 19 2486 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800008759.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000721.hg.1

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000399018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 ENST00000522917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800008759.hg.1 NM_001039112.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0