This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03927526_s1
See other SLC26A4-AS1 GE Assays ›
Gene Symbol:
SLC26A4-AS1
Gene Name:
SLC26A4 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 107656516 - 107661798 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03927526_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC26A4-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC052342.1 - 1 1251 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream B-cell receptor-associated protein 29 BCAP29 BAP29 -36.4kb Anti-sense Chr. 7 - 107579977 - 107623317 on NCBI Build GRCh38
Downstream Cbl proto-oncogene like 1 CBLL1 HAKAI,
RNF188
81.8kb Anti-sense Chr. 7 - 107743497 - 107764997 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000440512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000449741.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000587899.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000591896.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000626388.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000627161.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000629014.2 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 ENST00000630476.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Clariom D Assay TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Clariom D Assay TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Clariom D Assay TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Clariom D Assay TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Clariom D Assay TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Clariom D Assay TC0700012214.hg.1 NR_028137.1 Refseq
Affymetrix Human Clariom D Assay TC0700012214.hg.1 NR_028137.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 229842 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0