This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03917965_s1
See other LOC100128993 GE Assays ›
Gene Symbol:
LOC100128993
Gene Name:
uncharacterized LOC100128993
Gene Aliases:
-
Chromosome Location:
Chr.8: 19183674 - 19245522 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03917965_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100128993
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC033547.1 - 1 546 85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG723262.1 - 551 Chr. - - [-] on Build 85
GenBankmRNA HY224716.1 - 1 Chr. 8 - 19244855 - 19245317 [+] on Build GRCh38 85
GenBankmRNA HY206895.1 - 1 Chr. 8 - 19244850 - 19245341 [+] on Build GRCh38 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream pleckstrin and Sec7 domain containing 3 PSD3 EFA6D,
EFA6R,
HCA67
-157.9kb Sense Chr. 8 - 18527303 - 19086909 on NCBI Build GRCh38
Downstream SH2 domain containing 4A SH2D4A PPP1R38,
SH2A
67.9kb Anti-sense Chr. 8 - 19313443 - 19396218 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000517949.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000518417.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 ENST00000520920.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Clariom D Assay TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Clariom D Assay TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Clariom D Assay TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Clariom D Assay TC0800009774.hg.1 NR_038919.1 Refseq
Affymetrix Human Clariom D Assay TC0800009774.hg.1 NR_038919.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0