This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03906903_s1
See other PLXDC2 GE Assays ›
Gene Symbol:
PLXDC2
Gene Name:
plexin domain containing 2
Gene Aliases:
TEM7R
Chromosome Location:
Chr.10: 19816215 - 20289856 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03906903_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLXDC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282736.1 NP_001269665.1 13 5024 98
NM_032812.8 NP_116201.7 14 5171 98
GenBank mRNA AK127644.1 - 1 1748 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MAM and LDL receptor class A domain containing 1 MALRD1 C10orf112,
DIET1,
bA265G8.2
-546.5kb Sense Chr. 10 - 19046931 - 19734478 on NCBI Build GRCh38
Downstream microRNA 4675 MIR4675 269.0kb Sense Chr. 10 - 20551970 - 20552046 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLXDC2

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 137978 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0