This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03904619_s1
See other RCOR3 GE Assays ›
Gene Symbol:
RCOR3
Gene Name:
REST corepressor 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 211259347 - 211316385 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03904619_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RCOR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK057163.1 - 1 1550 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream potassium voltage-gated channel subfamily H member 1 KCNH1 EAG,
EAG1,
Kv10.1,
TMBTS,
ZLS1,
h-eag,
hEAG1
-157.1kb Anti-sense Chr. 1 - 210678314 - 211134115 on NCBI Build GRCh38
Downstream TNF receptor associated factor 5 TRAF5 MGC:39780,
RNF84
33.5kb Sense Chr. 1 - 211326587 - 211374946 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000487643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000487643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000487643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000487643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000341319.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000468734.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000487643.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000487643.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000487643.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000487643.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 ENST00000489245.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq
Affymetrix Human Clariom D Assay TC0300013344.hg.1 NM_130446.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header