This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03888517_m1
See other LOC105376010 GE Assays ›
Gene Symbol:
LOC105376010
Gene Name:
uncharacterized LOC105376010
Gene Aliases:
-
Chromosome Location:
Chr.9: 31578604 - 31744792 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03888517_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105376010
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK124280.1 - 4 - 5 327 84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB185943.1 - 4 - 5 327 Chr. 9 - 31578604 - 31738798 [+] on Build GRCh38 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Downstream aconitase 1 ACO1 ACONS,
HEL60,
IREB1,
IREBP,
IREBP1,
IRP1
650.7kb Sense Chr. 9 - 32384603 - 32450836 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700009169.hg.1 ENST00000574472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009169.hg.1 ENST00000574472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009169.hg.1 ENST00000574472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000615571.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000615571.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000616427.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000616427.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000618969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000618969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000585946.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000591521.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000615571.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000615571.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000616427.3 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000616427.3 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000618969.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000618969.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626130.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000626605.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014347.hg.1 ENST00000629837.2 Ensembl
Affymetrix Human Clariom D Assay TC1700009169.hg.1 ENST00000574472.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009169.hg.1 ENST00000574472.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009169.hg.1 ENST00000574472.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0