This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03870683_s1
See other GUCY1A2 GE Assays ›
Gene Symbol:
GUCY1A2
Gene Name:
guanylate cyclase 1 soluble subunit alpha 2
Gene Aliases:
GC-SA2, GUC1A2
Chromosome Location:
Chr.11: 106674012 - 107018445 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03870683_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GUCY1A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000855.2 NP_000846.1 8 5504 87
NM_001256424.1 NP_001243353.1 9 5597 87
GenBank mRNA AK098142.1 - 1 2155 87
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE645219.1 - 1 Chr. 11 - 106684423 - 106684850 [+] on Build GRCh38 87
GenBankmRNA AI560889.1 - 1 Chr. 11 - 106684425 - 106684923 [+] on Build GRCh38 87
GenBankmRNA DB304627.1 - 1 Chr. 11 - 106684425 - 106684988 [+] on Build GRCh38 87
GenBankmRNA BX110308.1 - 1 Chr. 11 - 106684418 - 106684848 [+] on Build GRCh38 87
GenBankmRNA AI091501.1 - 1 Chr. 11 - 106684422 - 106684809 [+] on Build GRCh38 87
GenBankmRNA DB348362.1 - 1 Chr. 11 - 106684425 - 106684961 [+] on Build GRCh38 87

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101928535 LOC101928535 -419.5kb Anti-sense Chr. 11 - 106250019 - 106264905 on NCBI Build GRCh38
Downstream uncharacterized LOC105369477 LOC105369477 586.5kb Sense Chr. 11 - 107223216 - 107318674 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002258.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GUCY1A2

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
signal transduction
positive regulation of cGMP biosynthetic process
intracellular signal transduction
adenylate cyclase activity
guanylate cyclase activity
GTP binding
heme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11026988.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11026988.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11026988.hg.1 TC11002258.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0