This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03859417_s1
See other RAB3C GE Assays ›
Gene Symbol:
RAB3C
Gene Name:
RAB3C, member RAS oncogene family
Gene Aliases:
-
Chromosome Location:
Chr.5: 58582188 - 58859394 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03859417_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAB3C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317915.1 NP_001304844.1 5 6646 106
NM_138453.3 NP_612462.1 5 6608 106
GenBank mRNA AK094685.1 - 1 655 106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA351349.1 - 1 187 Chr. 5 - 58856924 - 58857516 [+] on Build GRCh38 106
GenBankmRNA DA172544.1 - 1 394 Chr. 5 - 58856718 - 58857282 [+] on Build GRCh38 106
GenBankmRNA DA161108.1 - 1 300 Chr. 5 - 58856813 - 58857383 [+] on Build GRCh38 106
GenBankmRNA DA149156.1 - 1 81 Chr. 5 - 58857030 - 58857539 [+] on Build GRCh38 106
GenBankmRNA DA145740.1 - 1 352 Chr. 5 - 58856761 - 58857305 [+] on Build GRCh38 106
GenBankmRNA H05254.1 - 1 Chr. 5 - 58856966 - 58857332 [-] on Build GRCh38 106
GenBankmRNA DA304904.1 - 1 50 Chr. 5 - 58857061 - 58857631 [+] on Build GRCh38 106
GenBankmRNA DA290429.1 - 1 139 Chr. 5 - 58856972 - 58857547 [+] on Build GRCh38 106
GenBankmRNA AI685949.1 - 1 Chr. 5 - 58857032 - 58857327 [-] on Build GRCh38 106
GenBankmRNA DA707181.1 - 1 127 Chr. 5 - 58856984 - 58857558 [+] on Build GRCh38 106
GenBankmRNA BE645894.1 - 1 Chr. 5 - 58857032 - 58857329 [-] on Build GRCh38 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101928600 LOC101928600 -298.2kb Anti-sense Chr. 5 - 58541567 - 58558243 on NCBI Build GRCh38
Downstream phosphodiesterase 4D PDE4D ACRDYS2,
DPDE3,
HSPDE4D,
PDE43,
PDE4DN2,
STRK1
109.6kb Anti-sense Chr. 5 - 58969038 - 60488098 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RAB3C

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
protein transport
regulation of exocytosis
antigen processing and presentation
GTPase activity
protein binding
GTP binding
myosin V binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000381158.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000381158.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000381158.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000381158.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000513316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 ENST00000513316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000282878.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000381158.3 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000381158.3 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000381158.3 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000381158.3 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000507977.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000513316.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 ENST00000513316.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Clariom D Assay TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Clariom D Assay TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Clariom D Assay TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Clariom D Assay TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Clariom D Assay TC0500007494.hg.1 NM_138453.3 Refseq
Affymetrix Human Clariom D Assay TC0500007494.hg.1 NM_138453.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0