This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03855063_s1
See other LOC100288310 GE Assays ›
Gene Symbol:
LOC100288310
Gene Name:
uncharacterized LOC100288310
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03855063_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100288310
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB073392.1 - 1 1497 93
AK091686.1 - 1 1642 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream potassium voltage-gated channel subfamily B member 2 KCNB2 KV2.2 -9.8kb Sense Chr. 8 - 72537391 - 72945890 on NCBI Build GRCh38
Downstream telomeric repeat binding factor 1 TERF1 PIN2,
TRBF1,
TRF,
TRF1,
hTRF1-AS,
t-TRF1
58.4kb Sense Chr. 8 - 73008862 - 73047752 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495770 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495743 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495795 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495753 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495758 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495772 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495731 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212800 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212795 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212796 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212787 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495784 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495741 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212818 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495732 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495738 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212802 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212778 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212790 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212834 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212797 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212833 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495775 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495769 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495748 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212794 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212820 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495733 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495776 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495744 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16106123 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495774 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495788 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212765 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212822 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495785 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495751 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212779 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212789 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212803 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16106122 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495737 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495771 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212831 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212786 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495752 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495742 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495739 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16106121 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212759 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495740 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495747 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495791 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06212791 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26495773 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 569340 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18351931 - -
Agilent Human 1B Microarray G4111A A_32_P326216 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18351931 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800007948.hg.1 ENST00000564832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007948.hg.1 ENST00000564832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007948.hg.1 ENST00000596434.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007948.hg.1 ENST00000564832.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007948.hg.1 ENST00000564832.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007948.hg.1 ENST00000596434.1 Ensembl
Array Name Probe IDs Targets Target Type
Illumina human-6 v2.0 expression beadchip ILMN_1853196 - -
Illumina Sentrix Human-6 v2 Expression BeadChip 4860673 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_73244 - -
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1853196 - -
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1853196 - -
Illumina HumanWG-6_V2_0_R2 ILMN_1853196 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0