This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03853009_s1
See other KANSL1L GE Assays ›
Gene Symbol:
KANSL1L
Gene Name:
KAT8 regulatory NSL complex subunit 1 like
Gene Aliases:
C2orf67, MSL1v2
Chromosome Location:
Chr.2: 210021421 - 210171370 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03853009_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KANSL1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017003431.1 XP_016858920.1 1 3694 84
GenBank mRNA AK057810.1 - 1 1354 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101928020 LOC101928020 -15.5kb Anti-sense Chr. 2 - 210029922 - 210064356 on NCBI Build GRCh38
Downstream acyl-CoA dehydrogenase,
long chain
ACADL ACAD4,
LCAD
106.7kb Sense Chr. 2 - 210187990 - 210225491 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017003431.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000281772.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000415553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000415553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000415553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000415553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000415553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000415553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000415553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000418791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000428655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000429908.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000429908.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000429908.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000438563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000438563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000438563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000438563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000438563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000438563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000452086.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000453904.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000457374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000460256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000460256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000460256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 ENST00000460256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_001307976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015601.hg.1 NM_152519.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0