This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03847344_s1
See other LOC101928403 GE Assays ›
Gene Symbol:
LOC101928403
Gene Name:
uncharacterized LOC101928403
Gene Aliases:
-
Chromosome Location:
Chr.2: 88627829 - 88631825 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03847344_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101928403
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC017749.1 - 1 3247 106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX116270.1 - 1 Chr. 2 - 88631287 - 88631821 [-] on Build GRCh38 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream eukaryotic translation initiation factor 2 alpha kinase 3 EIF2AK3 PEK,
PERK,
WRS
-595.0 Anti-sense Chr. 2 - 88556740 - 88627576 on NCBI Build GRCh38
Downstream ribose 5-phosphate isomerase A RPIA RPI,
RPIAD
59.8kb Sense Chr. 2 - 88691658 - 88750935 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200008349.hg.1 ENST00000453008.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008349.hg.1 ENST00000453008.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008349.hg.1 ENST00000453008.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008349.hg.1 ENST00000453008.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008349.hg.1 ENST00000606164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008349.hg.1 ENST00000606164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008349.hg.1 ENST00000606164.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008349.hg.1 ENST00000453008.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008349.hg.1 ENST00000453008.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008349.hg.1 ENST00000453008.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008349.hg.1 ENST00000453008.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008349.hg.1 ENST00000606164.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008349.hg.1 ENST00000606164.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008349.hg.1 ENST00000606164.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0