This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03844183_s1
See other EPS15L1 GE Assays ›
Gene Symbol:
EPS15L1
Gene Name:
epidermal growth factor receptor pathway substrate 15 like 1
Gene Aliases:
EPS15R
Chromosome Location:
Chr.19: 16355244 - 16472012 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03844183_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPS15L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017027087.1 XP_016882576.1 25 2916 80
XM_017027088.1 XP_016882577.1 25 2809 80
GenBank mRNA BC028243.1 - 1 1553 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY312010.1 - 1 Chr. 19 - 16356578 - 16357012 [+] on Build GRCh38 80
GenBankmRNA R54896.1 - 61 Chr. - - [-] on Build 80
GenBankmRNA AW962695.1 - 1 302 Chr. 19 - 16356521 - 16357076 [-] on Build GRCh38 80
GenBankmRNA BI040124.1 - 1 165 Chr. 19 - 16356598 - 16356938 [-] on Build GRCh38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Kruppel like factor 2 KLF2 LKLF -28.8kb Anti-sense Chr. 19 - 16324826 - 16328662 on NCBI Build GRCh38
Downstream calreticulin 3 CALR3 CMH19,
CRT2,
CT93
120.7kb Sense Chr. 19 - 16479057 - 16496192 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
negative regulation of epidermal growth factor receptor signaling pathway
cell-cell adhesion
calcium ion binding
protein binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900009918.hg.1 ENST00000623994.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009918.hg.1 ENST00000623994.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0