This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03837329_s1
See other ATP6V0E1P2 GE Assays ›
Gene Symbol:
ATP6V0E1P2
Gene Name:
ATPase H+ transporting V0 subunit e1 pseudogene 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 41859154 - 41859641 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03837329_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP6V0E1P2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK022066.1 - 1 1950 117

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream unc-51 like kinase 4 ULK4 FAM7C1,
REC01035
Sense Chr. 3 - 41246599 - 41963020 on NCBI Build GRCh38
Downstream trafficking kinesin protein 1 TRAK1 MILT1,
OIP106
152.7kb Anti-sense Chr. 3 - 42013802 - 42225890 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008198.hg.1 ENST00000427294.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008198.hg.1 ENST00000427294.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010828.hg.1 ENST00000441771.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010828.hg.1 ENST00000441771.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008198.hg.1 ENST00000427294.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008198.hg.1 ENST00000427294.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0