This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03833905_s1
See other MAP3K14 GE Assays ›
Gene Symbol:
MAP3K14
Gene Name:
mitogen-activated protein kinase kinase kinase 14
Gene Aliases:
FTDCR1B, HS, HSNIK, NIK
Chromosome Location:
Chr.17: 45263119 - 45317064 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03833905_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAP3K14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AJ008141.1 - 1 57 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MAP3K14 antisense RNA 1 MAP3K14-AS1 -17.8kb Anti-sense Chr. 17 - 45247925 - 45268630 on NCBI Build GRCh38
Downstream Rho GTPase activating protein 27 ARHGAP27 CAMGAP1,
PP905,
SH3D20,
SH3P20
107.4kb Sense Chr. 17 - 45393902 - 45432916 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0