This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03833487_s1
See other RGS3 GE Assays ›
Gene Symbol:
RGS3
Gene Name:
regulator of G-protein signaling 3
Gene Aliases:
C2PA, RGP3
Chromosome Location:
Chr.9: 113444729 - 113597743 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03833487_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011518893.1 XP_011517195.1 21 5128 92
GenBank mRNA AF131806.1 - 1 1749 92
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ308278.1 - 1 Chr. 9 - 113543877 - 113544263 [+] on Build GRCh38 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chromosome 9 open reading frame 43 C9orf43 -112.8kb Sense Chr. 9 - 113410644 - 113429684 on NCBI Build GRCh38
Downstream zinc finger protein 618 ZNF618 FP13169,
NEDD10
332.0kb Sense Chr. 9 - 113876282 - 114056595 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

inactivation of MAPK activity
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
GTPase activator activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0