This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03832037_s1
See other EPS15 GE Assays ›
Gene Symbol:
EPS15
Gene Name:
epidermal growth factor receptor pathway substrate 15
Gene Aliases:
AF-1P, AF1P, MLLT5
Chromosome Location:
Chr.1: 51354263 - 51519354 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03832037_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPS15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017000615.1 XP_016856104.1 9 823 89
XM_017000616.1 XP_016856105.1 9 823 89
XM_017000620.1 XP_016856109.1 9 711 89
GenBank mRNA Z29070.1 - 1 49 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tetratricopeptide repeat domain 39A TTC39A C1orf34,
DEME-6
-102.8kb Sense Chr. 1 - 51287258 - 51345116 on NCBI Build GRCh38
Downstream oxysterol binding protein like 9 OSBPL9 ORP-9,
ORP9
168.7kb Anti-sense Chr. 1 - 51616874 - 51789219 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017000620.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

Golgi to endosome transport
epidermal growth factor receptor signaling pathway
cell proliferation
protein transport
vesicle organization
receptor-mediated endocytosis of virus by host cell
endocytic recycling
negative regulation of epidermal growth factor receptor signaling pathway
viral entry into host cell
clathrin coat assembly
cell-cell adhesion
calcium ion binding
protein binding
SH3 domain binding
polyubiquitin binding
identical protein binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0